ENA024293938 Enamine

[3-(morpholin-4-ylsulfonyl)phenyl]methanol

  • MDL number MFCD04635874
  • Cas Number : 646071-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07898

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07898
Availability details for
Quantity
  • Availability
  • Add to Cart