ENA024293945 Enamine

3-(3-bromophenyl)-3-(carbamoylamino)propanoic acid

  • MDL number MFCD05262778
  • Cas Number : 743440-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07905
Availability details for
Quantity
  • Availability
  • Add to Cart