ENA024293948 Enamine

3-(2-methylphenyl)-3-oxopropanenitrile

  • MDL number MFCD02260802
  • Cas Number : 35276-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...86
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07908

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-07908
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart