ENA024293950 Enamine

2-{2-[(4-chlorophenoxy)methyl]-1H-1,3-benzodiazol-1-yl}acetohydrazide

  • MDL number MFCD06335044
  • Cas Number : 750599-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07910
Availability details for
Quantity
  • Availability
  • Add to Cart