ENA024293953 Enamine

2-piperidin-4-yl-1,3-benzoxazole

  • MDL number MFCD01475844
  • Cas Number : 51784-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07913

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-07913
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart