ENA024293954 Enamine

4-(1,3-benzothiazol-2-yl)-2-chlorophenol

  • MDL number MFCD06336099
  • Cas Number : 749906-91-0
  • Molecular Weight : 261.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07914
Availability details for
Quantity
  • Availability
  • Add to Cart