ENA024293955 Enamine

4-(1,3-benzothiazol-2-yl)-2,6-dichlorophenol

  • MDL number MFCD04093460
  • Cas Number : 637302-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07915

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07915
Availability details for
Quantity
  • Availability
  • Add to Cart