ENA024293957 Enamine

3-{[3-(trifluoromethyl)phenyl]amino}-2,3-dihydro-1lambda6-thiophene-1,1-dione

  • MDL number MFCD02166494
  • Cas Number : 302948-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-144
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07917
Availability details for
Quantity
  • Availability
  • Add to Cart