ENA024293962 Enamine

3-(phenylamino)-2,3-dihydro-1lambda6-thiophene-1,1-dione

  • MDL number MFCD00188509
  • Cas Number : 39565-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07922
Availability details for
Quantity
  • Availability
  • Add to Cart