ENA024293967 Enamine

1-[(furan-2-yl)methyl]-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD04638748
  • Cas Number : 5049-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07927
Availability details for
Quantity
  • Availability
  • Add to Cart