ENA024293973 Enamine

4-({[(4-tert-butylphenyl)sulfonyl]amino}methyl)benzoic acid

  • MDL number MFCD02938130
  • Cas Number : 440350-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07933
Availability details for
Quantity
  • Availability
  • Add to Cart