ENA024293974 Enamine

4-(chloromethyl)-2-nitrophenol

  • MDL number MFCD02856711
  • Cas Number : 6694-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07934

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-07934
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart