ENA024293976 Enamine

2-chloro-N-ethyl-N-{[(propan-2-yl)carbamoyl]methyl}acetamide

  • MDL number MFCD04628564
  • Cas Number : 731821-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07936
Availability details for
Quantity
  • Availability
  • Add to Cart