ENA024293977 Enamine

3-(4-chloro-3-nitrobenzenesulfonamido)benzoic acid

  • MDL number MFCD04638744
  • Cas Number : 136832-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07937
Availability details for
Quantity
  • Availability
  • Add to Cart