ENA024293993 Enamine

2-(furan-2-yl)-2-(piperidin-1-yl)ethan-1-amine

  • MDL number MFCD06655011
  • Cas Number : 110358-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07953
Availability details for
Quantity
  • Availability
  • Add to Cart