ENA024293995 Enamine

methyl 5-bromo-2-(chlorosulfonyl)benzoate

  • MDL number MFCD04638740
  • Cas Number : 736948-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07955

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-07955
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart