ENA024294006 Enamine

N-[2-(4-chlorophenoxy)ethyl]-N-methylamine

  • MDL number MFCD00018645
  • Cas Number : 65686-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07966

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07966
Availability details for
Quantity
  • Availability
  • Add to Cart