ENA024294009 Enamine

3-amino-N,N-diethyl-4-[5-methyl-2-(propan-2-yl)phenoxy]benzene-1-sulfonamide

  • MDL number MFCD04638768
  • Cas Number : 736948-96-2
  • Molecular Weight : 376.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07969
Availability details for
Quantity
  • Availability
  • Add to Cart