ENA024294024 Enamine

2-{[4-ethyl-5-(2-methylphenyl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD03429376
  • Cas Number : 741731-80-6
  • Molecular Weight : 277.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07984
Availability details for
Quantity
  • Availability
  • Add to Cart