ENA024294025 Enamine

N-[(4-methylphenyl)methylidene]hydroxylamine

  • Cas Number : 3717-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07985
Availability details for
Quantity
  • Availability
  • Add to Cart