ENA024294037 Enamine

N-[1-(adamantan-1-yl)ethylidene]hydroxylamine

  • Cas Number : 78679-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07997
Availability details for
Quantity
  • Availability
  • Add to Cart