ENA024294044 Enamine

3-(1H-1,2,3,4-tetrazol-1-yl)phenol

  • MDL number MFCD01860802
  • Cas Number : 125620-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08004
Availability details for
Quantity
  • Availability
  • Add to Cart