ENA024294056 Enamine

2-chloro-N-[1-(4-fluorophenyl)ethyl]propanamide

  • MDL number MFCD06335040
  • Cas Number : 750599-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08016
Availability details for
Quantity
  • Availability
  • Add to Cart