ENA024294069 Enamine

1-amino-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid

  • MDL number MFCD00091959
  • Cas Number : 82-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08029
Availability details for
Quantity
  • Availability
  • Add to Cart