ENA024294077 Enamine

[(4-aminophenyl)sulfanyl]formonitrile

  • MDL number MFCD00042820
  • Cas Number : 2987-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-49
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08037
Availability details for
Quantity
  • Availability
  • Add to Cart