ENA024294096 Enamine

2-methyl-5-(piperidine-1-sulfonyl)benzoic acid

  • MDL number MFCD01417119
  • Cas Number : 329908-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08056
Availability details for
Quantity
  • Availability
  • Add to Cart