ENA024294119 Enamine

3-methyl-1-phenyl-5-(piperidin-1-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD01124432
  • Cas Number : 5499-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08079
Availability details for
Quantity
  • Availability
  • Add to Cart