ENA024294123 Enamine

1-amino-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00086187
  • Cas Number : 1562-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08083

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08083
Availability details for
Quantity
  • Availability
  • Add to Cart