ENA024294129 Enamine

1-(2-methoxyphenyl)-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD00834221
  • Cas Number : 15182-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08089
Availability details for
Quantity
  • Availability
  • Add to Cart