ENA024294140 Enamine

4-hydrazinyl-5-phenylthieno[2,3-d]pyrimidine

  • MDL number MFCD01763504
  • Cas Number : 113246-86-9
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 180-181
Iupac Name 4-hydrazino-5-phenylthieno[2,3-d]pyrimidine
Inchi Code 1S/C12H10N4S/c13-16-11-10-9(8-4-2-1-3-5-8)6-17-12(10)15-7-14-11/h1-7H,13H2,(H,14,15,16)
Inchi Key HKYLLTSMUHTLBT-UHFFFAOYSA-N

Description

Melting Point 180-181
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08100

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-08100
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart