ENA024294145 Enamine

2-chloro-1-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)ethan-1-one

  • MDL number MFCD01910939
  • Cas Number : 304685-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-100
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08105
Availability details for
Quantity
  • Availability
  • Add to Cart