ENA024294150 Enamine

3-pyridin-3-yl-2-thioxo-1,3-thiazolidin-4-one

  • MDL number MFCD01847264
  • Cas Number : 99419-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08110

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08110
Availability details for
Quantity
  • Availability
  • Add to Cart