ENA024294152 Enamine

adamantane-1-carbohydrazide

  • Cas Number : 17846-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08112
Availability details for
Quantity
  • Availability
  • Add to Cart