ENA024294157 Enamine

N,N-diallyl-2-chloroacetamide

  • MDL number MFCD00034186
  • Cas Number : 93-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08117

Safety Information

signalword Warning
safety_statement 26-28-36/37/39-61
msds https://www.enaminestore.com/catalog/EN300-08117
pictograms GHS07,GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart