ENA024294160 Enamine

2-chloro-N-(2-cyanoethyl)-N-(2-fluorophenyl)acetamide

  • MDL number MFCD03980971
  • Cas Number : 571149-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08120
Availability details for
Quantity
  • Availability
  • Add to Cart