ENA024294175 Enamine

N'-(chloroacetyl)-2-fluorobenzohydrazide

  • MDL number MFCD05262931
  • Cas Number : 499129-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08135

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08135
Availability details for
Quantity
  • Availability
  • Add to Cart