ENA024294182 Enamine

2-[(2-fluorophenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD04621499
  • Cas Number : 57978-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-216
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08142
Availability details for
Quantity
  • Availability
  • Add to Cart