ENA024294184 Enamine

5-(4-chlorophenyl)-4-(3-methoxypropyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03949049
  • Cas Number : 713097-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08144
Availability details for
Quantity
  • Availability
  • Add to Cart