ENA024294193 Enamine

2-chloro-N-[4-(2,4-dimethoxyphenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD04621711
  • Cas Number : 131213-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08153
Availability details for
Quantity
  • Availability
  • Add to Cart