ENA024294194 Enamine

2-chloro-N-[4-(2,5-dimethoxyphenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD04621532
  • Cas Number : 91402-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08154
Availability details for
Quantity
  • Availability
  • Add to Cart