ENA024294200 Enamine

4-hydroxy-1-(2-naphthylsulfonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD04619519

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08160

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08160
Availability details for
Quantity
  • Availability
  • Add to Cart