ENA024294219 Enamine

3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

  • MDL number MFCD00456558
  • Cas Number : 272437-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223...225
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08179

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08179
Availability details for
Quantity
  • Availability
  • Add to Cart