ENA024294220 Enamine

2-chloro-1-[1-(2-methoxyethyl)-2,5-dimethyl-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD03028474
  • Cas Number : 379241-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08180

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-08180
pictograms GHS05,GHS07
hstatements H302,H314,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart