ENA024294224 Enamine

2,4,5,7-tetraazatricyclo[6.4.0.0,2,6]dodeca-1(12),3,5,8,10-pentaene-3-thiol

  • MDL number MFCD01103518
  • Cas Number : 4290-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08184
Availability details for
Quantity
  • Availability
  • Add to Cart