ENA024294228 Enamine

4-(3-chloro-4-methylphenyl)-5-[(4-methoxyphenoxy)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03960512
  • Cas Number : 723332-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08188

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08188
Availability details for
Quantity
  • Availability
  • Add to Cart