ENA024294257 Enamine

3-methoxy-4-(2-oxo-2-phenylethoxy)benzaldehyde

  • MDL number MFCD03961133
  • Cas Number : 723332-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...129
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08217

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08217
Availability details for
Quantity
  • Availability
  • Add to Cart