ENA024294260 Enamine

4-[2-(3,4-dimethylphenyl)-2-oxoethoxy]benzaldehyde

  • MDL number MFCD03361599
  • Cas Number : 723332-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08220
Availability details for
Quantity
  • Availability
  • Add to Cart