ENA024294263 Enamine

2-chloro-1-{2,5-dimethyl-1-[(oxolan-2-yl)methyl]-1H-pyrrol-3-yl}ethan-1-one

  • MDL number MFCD03961617
  • Cas Number : 725710-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08223
Availability details for
Quantity
  • Availability
  • Add to Cart