ENA024294267 Enamine

3-nitro-4-(phenylthio)benzoic acid

  • MDL number MFCD01568962
  • Cas Number : 37531-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228...230
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08227

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08227
Availability details for
Quantity
  • Availability
  • Add to Cart