ENA024294275 Enamine

2-mercapto-3-(2-methoxybenzyl)quinazolin-4(3H)-one

  • MDL number MFCD02650871
  • Cas Number : 343618-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08235

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08235
Availability details for
Quantity
  • Availability
  • Add to Cart